Assay Design
Gene Transcripts
- Gene Symbol
- SH3BGRL2
- Entrez Gene ID
-
421851
- Gene Name
- SH3 domain binding glutamate rich protein like 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 79343244 - 79403585 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_419869.6 |
XP_419869.5 |
3 - 4 |
804 |
93 |
GenBank mRNA |
BX931923.1 |
- |
2 - 3 |
415 |
93 |
View Full Product Details