Assay Design
Gene Transcripts
- Gene Symbol
- TRAPPC3L
- Entrez Gene ID
-
421743
- Gene Name
- trafficking protein particle complex 3 like
- Gene Aliases
-
BET3L
- Chromosome Location
-
Chr.3: 63843807 - 63863465 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_419775.6 |
XP_419775.1 |
3 - 4 |
401 |
95 |
GenBank mRNA |
BX932116.1 |
- |
3 - 4 |
400 |
95 |
View Full Product Details