Assay Design



Gene Transcripts

Gene Symbol
PBLD
Entrez Gene ID
423685
Gene Name
phenazine biosynthesis like protein domain containing
Gene Aliases
-
Chromosome Location
Chr.6: 11263100 - 11281943 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277337.1 NP_001264266.1 1 - 2 271 110
GenBank mRNA BM487627.1 - 1 - 2 271 110
BX934609.1 - 1 - 2 149 110

View Full Product Details