Assay Design



Gene Transcripts

Gene Symbol
LRRC57
Entrez Gene ID
423236
Gene Name
leucine rich repeat containing 57
Gene Aliases
-
Chromosome Location
Chr.5: 25956738 - 25961495 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199668.1 NP_001186597.1 2 - 3 282 130
XM_015287078.2 XP_015142564.1 2 - 3 381 130
XM_015287079.2 XP_015142565.1 2 - 3 272 130
GenBank mRNA BU404676.1 - 2 - 3 282 130
BX934990.1 - 2 - 3 223 130
CN228613.1 - 2 - 3 201 130

View Full Product Details