Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276338.1 | NP_001263267.1 | 7 - 8 | 615 | 87 |
GenBank mRNA | BU204597.1 | - | 6 - 7 | 384 | 87 |
BU225566.1 | - | 7 - 8 | 88 | ||
BU324297.1 | - | 6 - 7 | 558 | 87 | |
BU328206.1 | - | 7 - 8 | 565 | 87 | |
BU328800.1 | - | 7 - 8 | 564 | 87 | |
CR352660.1 | - | 6 - 7 | 404 | 87 | |
CR352941.1 | - | 7 - 8 | 564 | 87 |