Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015279226.2 | XP_015134712.1 | 3 - 4 | 404 | 72 |
XM_015279233.1 | XP_015134719.1 | 3 - 4 | 429 | 72 | |
XM_025146857.1 | XP_025002625.1 | 3 - 4 | 712 | 72 | |
GenBank mRNA | CR353653.1 | - | 2 - 3 | 399 | 72 |