Assay Design



Gene Transcripts

Gene Symbol
SKA3
Entrez Gene ID
418948
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
C13orf3
Chromosome Location
Chr.1: 179931536 - 179940997 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015279226.2 XP_015134712.1 7 - 8 1318 109
XM_015279233.1 XP_015134719.1 7 - 8 1340 109
XM_025146857.1 XP_025002625.1 7 - 8 1626 109
GenBank mRNA CR353653.1 - 6 - 7 1313 109

View Full Product Details