Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015279226.2 | XP_015134712.1 | 8 - 9 | 1440 | 87 |
XM_015279233.1 | XP_015134719.1 | 8 - 9 | 1462 | 87 | |
XM_025146857.1 | XP_025002625.1 | 8 - 9 | 1748 | 87 | |
GenBank mRNA | CR353653.1 | - | 7 - 8 | 1435 | 87 |