Assay Design



Gene Transcripts

Gene Symbol
ACTBL2
Entrez Gene ID
426652
Gene Name
actin, beta-like 2
Gene Aliases
ACTBL2L, ACTC
Chromosome Location
Chr.5: 18553735 - 18572253 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025151131.1 XP_025006899.1 5 - 6 2014 106
XM_424279.6 XP_424279.1 6 - 7 2249 106
GenBank mRNA BX931992.2 - 5 - 6 646 106

View Full Product Details