Assay Design



Gene Transcripts

Gene Symbol
TUBAL3
Entrez Gene ID
416694
Gene Name
tubulin alpha like 3
Gene Aliases
TUBA3, TUBA3C
Chromosome Location
Chr.1: 1833939 - 1840449 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277351.1 NP_001264280.1 2 - 3 306 66
GenBank mRNA BU129060.1 - 1 - 2 257 66
BX935067.2 - 2 - 3 306 66

View Full Product Details