Assay Design



Gene Transcripts

Gene Symbol
MPZL3
Entrez Gene ID
419778
Gene Name
myelin protein zero like 3
Gene Aliases
-
Chromosome Location
Chr.24: 5465925 - 5471753 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252285.1 NP_001239214.1 2 - 3 286 66
GenBank mRNA CR353823.1 - 1 - 2 79 66

View Full Product Details