Assay Design



Gene Transcripts

Gene Symbol
APOPT1
Entrez Gene ID
423483
Gene Name
apoptogenic 1, mitochondrial
Gene Aliases
C14orf153, C5H14orf153
Chromosome Location
Chr.5: 50603347 - 50612237 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015287849.2 XP_015143335.1 3 - 4 115
GenBank mRNA BX929835.2 - 3 - 4 362 115
CR353689.1 - 2 - 3 477 115

View Full Product Details