Assay Design



Gene Transcripts

Gene Symbol
SYNDIG1
Entrez Gene ID
416717
Gene Name
synapse differentiation inducing 1
Gene Aliases
C20orf39, TMEM90B
Chromosome Location
Chr.3: 15797112 - 15861570 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015283540.2 XP_015139026.1 3 - 4 993 72
XM_015283541.2 XP_015139027.1 3 - 4 791 72
GenBank mRNA BX950498.2 - 4 - 5 697 72

View Full Product Details