Assay Design
Gene Transcripts
- Gene Symbol
- TMEM47
- Entrez Gene ID
-
418585
- Gene Name
- transmembrane protein 47
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 115288951 - 115315641 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_416786.6 |
XP_416786.1 |
2 - 3 |
484 |
68 |
GenBank mRNA |
CR388761.1 |
- |
2 - 3 |
468 |
68 |
View Full Product Details