Assay Design



Gene Transcripts

Gene Symbol
USPL1
Entrez Gene ID
395436
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
-
Chromosome Location
Chr.1: 176429802 - 176446252 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162393.2 NP_001155865.1 6 - 7 2387 129
XM_015277630.2 XP_015133116.1 7 - 8 1397 129
GenBank mRNA AF242552.1 - 4 - 5 513 129

View Full Product Details