Assay Design
Gene Transcripts
- Gene Symbol
- PFDN4
- Entrez Gene ID
-
419337
- Gene Name
- prefoldin subunit 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 12335201 - 12337124 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX930309.1 |
- |
1 - 2 |
32 |
74 |
View Full Product Details