Assay Design



Gene Transcripts

Gene Symbol
ELP6
Entrez Gene ID
420379
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
C2H3ORF75, C3orf75, TMEM103
Chromosome Location
Chr.2: 914076 - 929143 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277490.1 NP_001264419.1 3 - 4 223 71
GenBank mRNA BX932848.1 - 3 - 4 194 71

View Full Product Details