Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277557.3 | NP_001264486.1 | 4 - 5 | 456 | 127 |
XM_015290629.2 | XP_015146115.1 | 6 - 7 | 842 | 127 | |
XM_025152828.1 | XP_025008596.1 | 5 - 6 | 623 | 127 | |
XM_025152829.1 | XP_025008597.1 | 4 - 5 | 556 | 127 | |
XM_025152830.1 | XP_025008598.1 | 4 - 5 | 731 | 127 | |
XM_025152831.1 | XP_025008599.1 | 4 - 5 | 1157 | 127 | |
GenBank mRNA | BX934812.1 | - | 4 - 5 | 456 | 127 |