Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277557.3 | NP_001264486.1 | 6 - 7 | 624 | 126 |
XM_015290629.2 | XP_015146115.1 | 8 - 9 | 1010 | 126 | |
XM_025152828.1 | XP_025008596.1 | 7 - 8 | 791 | 126 | |
XM_025152829.1 | XP_025008597.1 | 6 - 7 | 724 | 126 | |
XM_025152830.1 | XP_025008598.1 | 6 - 7 | 899 | 126 | |
XM_025152831.1 | XP_025008599.1 | 6 - 7 | 1325 | 126 | |
GenBank mRNA | BX934812.1 | - | 6 - 7 | 624 | 126 |