Assay Design



Gene Transcripts

Gene Symbol
FGA
Entrez Gene ID
396307
Gene Name
fibrinogen alpha chain
Gene Aliases
alpha-fibrinogen
Chromosome Location
Chr.4: 20321300 - 20328191 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271911.1 NP_001258840.1 4 - 5 537 197
NM_205356.2 NP_990687.2 4 - 5 537 197
GenBank mRNA BX935039.1 - 4 - 5 519 94
M34096.1 - 4 - 5 522 197

View Full Product Details