Assay Design



Gene Transcripts

Gene Symbol
ZFP91
Entrez Gene ID
423128
Gene Name
ZFP91 ZFP91 zinc finger protein
Gene Aliases
ZFP91-CNTF
Chromosome Location
Chr.5: 17173776 - 17188554 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015286837.2 XP_015142323.2 10 - 11 1530 61
GenBank mRNA AJ851756.1 - 10 - 11 987 61

View Full Product Details