Assay Design



Gene Transcripts

Gene Symbol
PTCD1
Entrez Gene ID
416493
Gene Name
pentatricopeptide repeat domain 1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001369063.1 NP_001355992.1 2 - 3 635 82
GenBank mRNA CR390934.1 - 2 - 3 237 82

View Full Product Details