Assay Design



Gene Transcripts

Gene Symbol
TICRR
Entrez Gene ID
415489
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Gene Aliases
C10H15ORF42, C15orf42, SLD3, treslin
Chromosome Location
Chr.10: 13279027 - 13295655 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285869.1 NP_001272798.1 5 - 6 1607 139
XM_025153845.1 XP_025009613.1 5 - 6 1684 139
GenBank mRNA CR407392.1 - 1 - 2 128 139

View Full Product Details