Assay Design
Gene Transcripts
- Gene Symbol
- EMC6
- Entrez Gene ID
-
417604
- Gene Name
- ER membrane protein complex subunit 6
- Gene Aliases
-
TMEM93
- Chromosome Location
-
Chr.19: 6786466 - 6788568 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CR407509.1 |
- |
1 - 1 |
252 |
122 |
View Full Product Details