Assay Design
Gene Transcripts
- Gene Symbol
- BOD1
- Entrez Gene ID
-
416210
- Gene Name
- biorientation of chromosomes in cell division 1
- Gene Aliases
-
FAM44B
- Chromosome Location
-
Chr.13: 10056287 - 10061871 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_414535.6 |
XP_414535.3 |
2 - 3 |
575 |
88 |
GenBank mRNA |
CR523593.1 |
- |
1 - 2 |
161 |
88 |
View Full Product Details