Assay Design
Gene Transcripts
- Gene Symbol
- TCRA
- Entrez Gene ID
-
426841
- Gene Name
- T-cell receptor V alpha
- Gene Aliases
-
-
- Chromosome Location
-
Chr.27: 2606363 - 2612599 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX932894.1 |
- |
2 - 2 |
450 |
138 |
BX934270.2 |
- |
2 - 2 |
446 |
138 |
View Full Product Details