Assay Design



Gene Transcripts

Gene Symbol
MCFD2
Entrez Gene ID
421413
Gene Name
multiple coagulation factor deficiency 2
Gene Aliases
-
Chromosome Location
Chr.3: 27051311 - 27055352 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006400.1 NP_001006400.1 3 - 4 331 71
XM_015283700.2 XP_015139186.1 3 - 4 394 71
XM_025148682.1 XP_025004450.1 2 - 3 292 71
GenBank mRNA AJ720519.1 - 3 - 4 331 71

View Full Product Details