Assay Design



Gene Transcripts

Gene Symbol
EIF3M
Entrez Gene ID
421602
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
PCID1
Chromosome Location
Chr.5: 5765073 - 5779380 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006406.1 NP_001006406.1 9 - 10 998 97
GenBank mRNA AJ720570.1 - 9 - 10 998 97
CR352624.1 - 9 - 10 982 97

View Full Product Details