Assay Design



Gene Transcripts

Gene Symbol
CHMP1B
Entrez Gene ID
422143
Gene Name
charged multivesicular body protein 1B
Gene Aliases
-
Chromosome Location
Chr.4: 1522246 - 1528803 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006428.1 NP_001006428.1 5 - 6 502 70
GenBank mRNA AJ719963.1 - 5 - 6 502 70
CR387811.1 - 5 - 6 519 70

View Full Product Details