Assay Design
Gene Transcripts
- Gene Symbol
- NSG1
- Entrez Gene ID
-
395251
- Gene Name
- neuronal vesicle trafficking associated 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 78573919 - 78594504 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204565.1 |
NP_989896.1 |
4 - 5 |
397 |
115 |
GenBank mRNA |
AF498103.1 |
- |
4 - 5 |
397 |
115 |
View Full Product Details