Assay Design



Gene Transcripts

Gene Symbol
NR2E3
Entrez Gene ID
395289
Gene Name
nuclear receptor subfamily 2 group E member 3
Gene Aliases
PNR
Chromosome Location
Chr.10: 1852294 - 1857167 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204594.1 NP_989925.1 3 - 4 330 93
XM_025153796.1 XP_025009564.1 2 - 3 360 93
GenBank mRNA AY053465.1 - 3 - 4 330 93

View Full Product Details