Assay Design



Gene Transcripts

Gene Symbol
GNRHR
Entrez Gene ID
395368
Gene Name
gonadotropin releasing hormone receptor
Gene Aliases
CGNRH-R, CGNRHRL
Chromosome Location
Chr.10: 18239128 - 18254320 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204653.1 NP_989984.1 2 - 3 710 83
XM_015292070.2 XP_015147556.1 4 - 5 4176 83
GenBank mRNA AJ304414.1 - 2 - 3 710 83

View Full Product Details