Assay Design
Gene Transcripts
- Gene Symbol
- ATOH7
- Entrez Gene ID
-
395388
- Gene Name
- atonal bHLH transcription factor 7
- Gene Aliases
-
ATH5
- Chromosome Location
-
Chr.6: 11251530 - 11252043 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204668.1 |
NP_989999.1 |
1 - 1 |
453 |
67 |
GenBank mRNA |
AJ001178.1 |
- |
1 - 1 |
453 |
67 |
- Gene Symbol
- MYPN
- Entrez Gene ID
-
423684
- Gene Name
- myopalladin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 11173017 - 11252666 on Build GRCg6a
View Full Product Details