Assay Design



Gene Transcripts

Gene Symbol
ZP1
Entrez Gene ID
395418
Gene Name
zona pellucida glycoprotein 1
Gene Aliases
-
Chromosome Location
Chr.5: 264554 - 268948 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204683.2 NP_990014.2 8 - 9 2282 151
GenBank mRNA AJ289697.1 - 8 - 9 2282 151
LC128587.1 - 7 - 8 2353 151

View Full Product Details