Assay Design



Gene Transcripts

Gene Symbol
CFC1
Entrez Gene ID
395437
Gene Name
cripto, FRL-1, cryptic family 1
Gene Aliases
CFC1B
Chromosome Location
Chr.Z: 42432327 - 42436992 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204700.2 NP_990031.1 2 - 3 244 84
GenBank mRNA AF228760.1 - 2 - 3 245 84
AF282984.1 - 2 - 3 206 84

View Full Product Details