Assay Design



Gene Transcripts

Gene Symbol
NTM
Entrez Gene ID
395450
Gene Name
neurotrimin
Gene Aliases
CEPU, CEPU-SE, HNT
Chromosome Location
Chr.24: 1826279 - 2138596 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204711.1 NP_990042.1 8 - 9 1047 63
XM_015297952.2 XP_015153438.1 8 - 9 1322 63
XM_015297955.2 XP_015153441.1 9 - 10 1194 63
XM_015297956.2 XP_015153442.1 8 - 9 916 63
XM_025143176.1 XP_024998944.1 7 - 8 846 63
GenBank mRNA Z72497.1 - 8 - 9 1047 63

View Full Product Details