Assay Design



Gene Transcripts

Gene Symbol
SYNM
Entrez Gene ID
395599
Gene Name
synemin
Gene Aliases
DMN
Chromosome Location
Chr.10: 17303647 - 17324081 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204809.4 NP_990140.2 1 - 2 862 69

View Full Product Details