Assay Design
Gene Transcripts
- Gene Symbol
- CHRNB4
- Entrez Gene ID
-
395613
- Gene Name
- cholinergic receptor nicotinic beta 4 subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 4055642 - 4067291 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204819.1 |
NP_990150.1 |
1 - 2 |
202 |
75 |
GenBank mRNA |
J05643.2 |
- |
1 - 2 |
202 |
75 |
View Full Product Details