Assay Design
Gene Transcripts
- Gene Symbol
- RLIM
- Entrez Gene ID
-
395630
- Gene Name
- ring finger protein, LIM domain interacting
- Gene Aliases
-
R-LIM, RNF12
- Chromosome Location
-
Chr.4: 12338165 - 12356466 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204828.1 |
NP_990159.1 |
2 - 3 |
310 |
76 |
GenBank mRNA |
AF069993.1 |
- |
2 - 3 |
310 |
76 |
View Full Product Details