Assay Design



Gene Transcripts

Gene Symbol
RPS6
Entrez Gene ID
396148
Gene Name
ribosomal protein S6
Gene Aliases
-
Chromosome Location
Chr.Z: 33736418 - 33740876 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205225.2 NP_990556.1 4 - 5 515 81
GenBank mRNA AF208228.1 - 3 - 4 493 81
X81968.1 - - 516 81

View Full Product Details