Assay Design



Gene Transcripts

Gene Symbol
KBP
Entrez Gene ID
396300
Gene Name
kainate binding protein
Gene Aliases
-
Chromosome Location
Chr.21: 1456666 - 1471680 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205353.2 NP_990684.1 5 - 6 462 110
XM_025142488.1 XP_024998256.1 5 - 6 1283 110
GenBank mRNA BI394598.1 - 5 - 6 462 110
X17700.1 - - 454 110

View Full Product Details