Assay Design



Gene Transcripts

Gene Symbol
VIP
Entrez Gene ID
396323
Gene Name
vasoactive intestinal peptide
Gene Aliases
-
Chromosome Location
Chr.3: 49586424 - 49593144 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177309.1 NP_001170780.1 5 - 6 672 121
NM_205366.2 NP_990697.2 6 - 7 777 121
GenBank mRNA BU391863.1 - 3 - 4 374 121
BU398589.1 - 4 - 5 507 121
U09350.1 - 6 - 7 753 121
X80906.1 - - 613 121

View Full Product Details