Assay Design
Gene Transcripts
- Gene Symbol
- SDC3
- Entrez Gene ID
-
396343
- Gene Name
- syndecan 3
- Gene Aliases
-
syndecan-3
- Chromosome Location
-
Chr.23: 550252 - 568556 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205383.1 |
NP_990714.1 |
2 - 3 |
215 |
76 |
GenBank mRNA |
M84910.1 |
- |
2 - 3 |
215 |
76 |
View Full Product Details