Assay Design
Gene Transcripts
- Gene Symbol
- ANOS1
- Entrez Gene ID
-
396395
- Gene Name
- anosmin 1
- Gene Aliases
-
KAL, KAL1, anosmin-1
- Chromosome Location
-
Chr.1: 127043959 - 127168215 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205424.1 |
NP_990755.1 |
1 - 2 |
191 |
58 |
GenBank mRNA |
L12144.1 |
- |
1 - 2 |
191 |
58 |
View Full Product Details