Assay Design



Gene Transcripts

Gene Symbol
ANOS1
Entrez Gene ID
396395
Gene Name
anosmin 1
Gene Aliases
KAL, KAL1, anosmin-1
Chromosome Location
Chr.1: 127043959 - 127168215 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205424.1 NP_990755.1 6 - 7 837 151
GenBank mRNA CR386644.1 - 3 - 4 360 151
L12144.1 - 6 - 7 837 151

View Full Product Details