Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_205486.1 | NP_990817.1 | 2 - 3 | 229 | 66 |
XM_025149536.1 | XP_025005304.1 | 1 - 2 | 172 | 66 | |
GenBank mRNA | BX935920.2 | - | 2 - 3 | 238 | 66 |
M80574.1 | - | 2 - 3 | 246 | 66 | |
M83235.1 | - | 2 - 3 | 229 | 66 |