Assay Design



Gene Transcripts

Gene Symbol
GLUL
Entrez Gene ID
396489
Gene Name
glutamate-ammonia ligase
Gene Aliases
p42
Chromosome Location
Chr.8: 6016293 - 6022834 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205493.1 NP_990824.1 5 - 6 700 95
GenBank mRNA M29076.1 - 5 - 6 700 95
S45408.1 - 4 - 5 660 95

View Full Product Details