Assay Design
Gene Transcripts
- Gene Symbol
- CAPZA1
- Entrez Gene ID
-
396521
- Gene Name
- capping actin protein of muscle Z-line alpha subunit 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.26: 3533331 - 3543277 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205515.1 |
NP_990846.1 |
- |
904 |
98 |
GenBank mRNA |
M25534.1 |
- |
- |
904 |
98 |
View Full Product Details