Assay Design



Gene Transcripts

Gene Symbol
TRA2A
Entrez Gene ID
420618
Gene Name
transformer 2 alpha homolog
Gene Aliases
-
Chromosome Location
Chr.2: 31280845 - 31304603 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006360.1 NP_001006360.1 6 - 7 937 114
XM_025147197.1 XP_025002965.1 7 - 8 1235 114
GenBank mRNA AJ719598.1 - 6 - 7 937 114

View Full Product Details