Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012318.1 | NP_001012318.1 | 3 - 4 | 374 | 96 |
GenBank mRNA | AF322051.1 | - | 3 - 4 | 358 | 96 |
AF377322.1 | - | 2 - 3 | 240 | 96 | |
BU135446.1 | - | 3 - 4 | 337 | 96 | |
BX275001.4 | - | 1 - 2 | 64 | 96 | |
CN227820.1 | - | 3 - 4 | 374 | 96 |